2,4,6-Trimethylbenzylamine, 99%
2,4,6-Trimethylbenzylamine, 99%
2,4,6-Trimethylbenzylamine, 99%
Thermo Scientific Chemicals

2,4,6-Trimethylbenzylamine, 99%

CAS: 40393-99-5 | C10H16N | 150.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 351180050
Price (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS40393-99-5
IUPAC Name(2,4,6-trimethylphenyl)methanaminium
Molecular FormulaC10H16N
InChI KeyDGSRAILDFBJNQI-UHFFFAOYSA-O
SMILESCC1=CC(C)=C(C[NH3+])C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5360 to 1.5380 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only