Tetra-n-butylammonium hexafluorophosphate, 98%
Tetra-n-butylammonium hexafluorophosphate, 98%
Tetra-n-butylammonium hexafluorophosphate, 98%
Thermo Scientific Chemicals

Tetra-n-butylammonium hexafluorophosphate, 98%

CAS: 3109-63-5 | C16H36F6NP | 387.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 352585000
Price (USD)
816.65
Special offer
Online exclusive
Ends: 28-Dec-2026
961.00
Save 144.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
816.65
Special offer
Online exclusive
Ends: 28-Dec-2026
961.00
Save 144.35
Each
Chemical Identifiers
CAS3109-63-5
IUPAC Namehexafluoro-λ⁵-phosphanuide; tetrabutylazanium
Molecular FormulaC16H36F6NP
InChI KeyBKBKEFQIOUYLBC-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder and/or chunks
Elemental Analysis>=97.5 %
Melting point242°C to 247°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only