3-Aminophenylboronic acid monohydrate, 98%
3-Aminophenylboronic acid monohydrate, 98%
3-Aminophenylboronic acid monohydrate, 98%
Thermo Scientific Chemicals

3-Aminophenylboronic acid monohydrate, 98%

CAS: 206658-89-1 | C6H10BNO3 | 154.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 352760050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS206658-89-1
IUPAC Name(3-aminophenyl)boronic acid hydrate
Molecular FormulaC6H10BNO3
InChI KeyXAEOVQODHLLNKX-UHFFFAOYSA-N
SMILESO.NC1=CC=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to brown
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only