Methyl phenyl sulfone, 99%
Methyl phenyl sulfone, 99%
Methyl phenyl sulfone, 99%
Thermo Scientific Chemicals

Methyl phenyl sulfone, 99%

CAS: 3112-85-4 | C7H8O2S | 156.2 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 353040250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Assay Percent Range98.5% min. (GC)
CAS3112-85-4
Chemical Name or MaterialMethyl phenyl sulfone
ColorWhite
EINECS Number221-476-8
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only