Thermo Scientific Chemicals

Tetrabutylammonium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Catalog number: 353090050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

Tetrabutylammonium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Catalog number: 353090050
5 g, Glass Bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
429-42-5
IUPAC Name
tetrabutylazanium; tetrafluoroboranuide
Molecular Formula
C16H36BF4N
InChI Key
NNZZSJSQYOFZAM-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
Assay
>=97.5 %
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text