1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%
Thermo Scientific Chemicals

1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%

CAS: 304680-35-1 | C10H19F6N2P | 312.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number 354181000
Price (USD)
721.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
721.00
Each
Chemical Identifiers
CAS304680-35-1
IUPAC Name1-hexyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC10H19F6N2P
InChI KeyYPWSSSRXUOQNMQ-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
NMR>=98.5 %
Infrared spectrumConforms
Refractive index1.4170 to 1.4200 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquid
RUO – Research Use Only