1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
Thermo Scientific Chemicals

1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%

CAS: 174501-65-6 | C8H15BF4N2 | 226.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 354210250
Price (USD)
130.65
Online exclusive
145.00
Save 14.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
130.65
Online exclusive
145.00
Save 14.35
Each
Chemical Identifiers
CAS174501-65-6
IUPAC Name1-butyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Molecular FormulaC8H15BF4N2
InChI KeyLSBXQLQATZTAPE-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange
Appearance (Form)Viscous liquid
Infrared spectrumConforms
HPLC>=98.0 %
Water=<1.0 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquid
RUO – Research Use Only