TentaGel™ S-NH2
TentaGel™ S-NH2
TentaGel™ S-NH2
Thermo Scientific Chemicals

TentaGel™ S-NH2

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Glass Bottle
Catalog number 354610250
Price (USD)
742.65
Special offer
Online exclusive
Ends: 28-Dec-2026
874.00
Save 131.35
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
742.65
Special offer
Online exclusive
Ends: 28-Dec-2026
874.00
Save 131.35
Each
Specifications
Chemical Name or MaterialTentaGel™ S-NH2
ColorWhite to Yellow
Loading0.2 to 0.3mmol/g
MDL NumberMFCD00217899
PackagingGlass Bottle
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesis
RUO – Research Use Only