TentaGel™ S-RAM
TentaGel™ S-RAM
TentaGel™ S-RAM
Thermo Scientific Chemicals

TentaGel™ S-RAM

 
Have Questions?
Catalog NumberQuantityPackaging
3546500505 gGlass bottle
Catalog number 354650050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTentaGel™ S-RAM
ColorWhite to Yellow
Loading0.2 to 0.3mmol/g
MDL NumberMFCD00217896
PackagingGlass bottle
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesis
RUO – Research Use Only