Nunc™ Biobanking and Cell Culture Cryogenic Tube Accessories, blue coder - FAQs
Nunc™ Biobanking and Cell Culture Cryogenic Tube Accessories, blue coder - FAQs
View additional product information for Cryo Vial Closure Color Coders - FAQs (354879, 375930, 375884PK, 354755, 355077, 354968PK, 355158PK, 375930PK, 354755PK, 354968, 375884, 375906, 355158, 355018PK, 355077PK, 355018, 375868, 5045-0005PK, 375922, 5045-0004PK, 5045-0003PK, 5045-0002PK, 354879PK, 5045-0000PK, 375906PK, 375868PK, 5045-0000, 5045-0002, 5045-0003, 375922PK, 5045-0004, 5045-0005)
1 product FAQs found
Are your Nunc CryoColor Vial Closure Coders removable?
Once inserted, the Nunc CryoColor Vial Closure Coders cannot be removed.