Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Thermo Scientific™

Cryo Vial Closure Color Coders

Green features
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct LineUnit Size
354968RedNuncCase of 2000
354755WhiteNuncCase of 2000
354755PK
also known as 354755
WhiteNuncPack of 500
354879BlueNuncCase of 2000
354879PK
also known as 354879
BlueNuncPack of 500
354968PK
also known as 354968
RedNuncPack of 500
355018GreenNuncCase of 2000
355018PK
also known as 355018
GreenNuncPack of 500
355077YellowNuncCase of 2000
355077PK
also known as 355077
YellowNuncPack of 500
355158OrangeNuncCase of 2000
355158PK
also known as 355158
OrangeNuncPack of 500
375868BrownNuncCase of 2000
375868PK
also known as 375868
BrownNuncPack of 500
375884PinkNuncCase of 2000
375884PK
also known as 375884
PinkNuncPack of 500
375906GrayNuncCase of 2000
375906PK
also known as 375906
GrayNuncPack of 500
375922PurpleNuncCase of 2000
375922PK
also known as 375922
PurpleNuncPack of 500
375930AssortedNuncCase of 2000
375930PK
also known as 375930
AssortedNuncPack of 500
5045-0003BlueNalgeneCase of 500
5045-0003PK
also known as 5045-0003
BlueNalgenePack of 100
5045-0000WhiteNalgeneCase of 500
5045-0000PK
also known as 5045-0000
WhiteNalgenePack of 100
5045-0002YellowNalgeneCase of 500
5045-0002PK
also known as 5045-0002
YellowNalgenePack of 100
5045-0004GreenNalgeneCase of 500
5045-0004PK
also known as 5045-0004
GreenNalgenePack of 100
5045-0005RedNalgeneCase of 500
5045-0005PK
also known as 5045-0005
RedNalgenePack of 100
Catalog number 354968
Price (USD)
224.72
Online exclusive
261.87
Save 37.15
Case of 2000
Color:
Red
Product Line:
Nunc
Unit Size:
Case of 2000
Price (USD)
224.72
Online exclusive
261.87
Save 37.15
Case of 2000
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.
  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
DescriptionNunc™ CryoColor Vial Closure Coder, PS
For Use With (Equipment)Cryogenic Vial
Product TypeVial Closure
ColorRed
MaterialPolystyrene
Product LineNunc
Quantity500/Pk., 2000/Cs.
Unit SizeCase of 2000

Frequently asked questions (FAQs)

Are your Nunc CryoColor Vial Closure Coders removable?

Once inserted, the Nunc CryoColor Vial Closure Coders cannot be removed.