n-Octadecyl mercaptan, 96%
n-Octadecyl mercaptan, 96%
n-Octadecyl mercaptan, 96%
Thermo Scientific Chemicals

n-Octadecyl mercaptan, 96%

CAS: 2885-00-9 | C18H38S | 286.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 354980250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2885-00-9
IUPAC Nameoctadecane-1-thiol
Molecular FormulaC18H38S
InChI KeyQJAOYSPHSNGHNC-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point25°C to 35°C
Appearance (Form)Crystalline solid
GC>=95 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only