4-(Trifluoromethyl)thiophenol, 97%
4-(Trifluoromethyl)thiophenol, 97%
4-(Trifluoromethyl)thiophenol, 97%
Thermo Scientific Chemicals

4-(Trifluoromethyl)thiophenol, 97%

CAS: 825-83-2 | C7H5F3S | 178.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3552900101 gGlass bottle
Catalog number 355290010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS825-83-2
IUPAC Name[4-(trifluoromethyl)phenyl]sulfanide
Molecular FormulaC7H4F3S
InChI KeyWCMLRSZJUIKVCW-UHFFFAOYSA-M
SMILESFC(F)(F)C1=CC=C([S-])C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4920 to 1.4950 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only