Tris(4-trifluoromethylphenyl)phosphine, 99%
Tris(4-trifluoromethylphenyl)phosphine, 99%
Tris(4-trifluoromethylphenyl)phosphine, 99%
Thermo Scientific Chemicals

Tris(4-trifluoromethylphenyl)phosphine, 99%

CAS: 13406-29-6 | C21H12F9P | 466.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 355340010
Price (USD)
129.65
Special offer
Online exclusive
Ends: 28-Dec-2026
152.00
Save 22.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
129.65
Special offer
Online exclusive
Ends: 28-Dec-2026
152.00
Save 22.35
Each
Chemical Identifiers
CAS13406-29-6
IUPAC Nametris[4-(trifluoromethyl)phenyl]phosphane
Molecular FormulaC21H12F9P
InChI KeyPXYCJKZSCDFXLR-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(C=C1)P(C1=CC=C(C=C1)C(F)(F)F)C1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Off-white to light yellow
GC>=98.5 %
Infrared spectrumConforms
Melting point70°C to 75°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only