Chloromethyl ethyl ether, 80%, Tech., stabilized
Chloromethyl ethyl ether, 80%, Tech., stabilized
Chloromethyl ethyl ether, 80%, Tech., stabilized
Thermo Scientific Chemicals

Chloromethyl ethyl ether, 80%, Tech., stabilized

CAS: 3188-13-4 | C3H7ClO | 94.54 g/mol
Have Questions?
Catalog NumberQuantityPackaging
35572025025 mLGlass bottle
Catalog number 355720250
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
91.50
Save 13.85
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
91.50
Save 13.85
Each
Chemical Identifiers
CAS3188-13-4
IUPAC Name(chloromethoxy)ethane
Molecular FormulaC3H7ClO
InChI KeyFCYRSDMGOLYDHL-UHFFFAOYSA-N
SMILESCCOCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=75 %
StabilizerCaCl2
Refractive index1.4000 to 1.4050 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only