(S)-(-)-2,2'-Diamino-1,1'-binaphthalene, 97%, Thermo Scientific Chemicals
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene, 97%, Thermo Scientific Chemicals
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(-)-2,2'-Diamino-1,1'-binaphthalene, 97%, Thermo Scientific Chemicals

CAS: 18531-95-8 | Molecular Formula: C20H16N2 | Molecular Weight: 284.36 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3565200101 gGlass bottle
Catalog number 356520010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS18531-95-8
IUPAC Name[1,1'-binaphthalene]-2,2'-diamine
Molecular FormulaC20H16N2
InChI KeyDDAPSNKEOHDLKB-UHFFFAOYSA-N
SMILESNC1=CC=C2C=CC=CC2=C1C1=C(N)C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO496.0 to 104.0 %
Appearance (Form)Powder
HPLC>=96.0 %
Specific optical rotation-149.5° to -155.5° (20°C, 589 nm) (c=1, pyridine)
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only