Silica gel, for drying purposes, non-toxic grade, 3-6 mm
Silica gel, for drying purposes, non-toxic grade, 3-6 mm
Silica gel, for drying purposes, non-toxic grade, 3-6 mm
Thermo Scientific Chemicals

Silica gel, for drying purposes, non-toxic grade, 3-6 mm

Silica gel, O2Si, CAS Number-7631-86-9, Silicon dioxide, 25kg, 2.0% max. (140 deg.C, 4 hrs), Adsorption capacity (relative humidity 40%): ca 20%, 620 to 800g/L, CHEBI:30563, White to Yellow, 231-545-4, 60.08, Non-toxic, VYPSYNLAJGMNEJ-UHFFFAOYSA-N, dioxosilane, 15, 8632, MFCD00011232 | CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Packaging:
View all
Plastic Bottle
Catalog number 357400010
Price (USD)
94.70
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
94.70
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Irregular granules
Particle size3.0 to 6.0 mm
Appearance (Color)Colorless to white
Residual water=<2.0 % (140°C, 4 h)
Water adsorption>=20.0 % (40% rH)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent, For column chromatography
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only