Cycloheximide, 95%
Cycloheximide, 95%
Cycloheximide, 95%
Thermo Scientific Chemicals

Cycloheximide, 95%

Cycloheximide, CAS # 66-81-9, is a compound produced by the bacterium Streptomyces griseus that shows an antifungal antibiotic activity with the ability to inhibit protein synthesis only in eukaryotic cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
3574200101g
3574200505g
35742025025g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 357420010
Price (USD)
94.50
Each
Quantity:
1g
Request bulk or custom format
Price (USD)
94.50
Each
Chemical Identifiers
CAS66-81-9
IUPAC Name4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
Molecular FormulaC15H23NO4
InChI KeyYPHMISFOHDHNIV-QTEFRXOUNA-N
SMILES[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Specific optical rotation-25° to -32° (20°C, 589 nm) (c=1, CHCl3)
Melting point108°C to 118°C
HPLC>=94 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cycloheximide is a semisynthetic compound showing an antifungal antibiotic activity.
  • It is produced by the bacterium Streptomyces griseus
  • Cycloheximide can inhibit protein synthesis in eukaryotic cells but not in prokaryotes by interacting directly with the translocase enzyme and interfering with the translocation step. Furthermore, this compound also blocks the A site of ribosomes

Application

  • Cycloheximide shows a neuroprotective, fungicide, and anticoronaviral activities
  • In biomedical research, it can induce glycogenolysis, gluconeogenesis, and ureagenesis
  • Due to significant toxic side effects such as teratogenesis, DNA damage, birth defects, and toxicity to sperm, this chemical compound is only used in in vitro research applications
  • This compound also shows inhibitory activity against ferroptosis
  • It is also used in laboratory media as a selective agent to permit the isolation of pathogenic and non-pathogenic fungi from other types of specimens
  • In laboratory research related to plant biology, it can induce ethylene production to promote the plant growth
  • In laboratory research, this compound can be applied to determine the half-life of protein in any cellular system. The treatment of in vitro cells with cycloheximide in a time-course experiment followed by western blotting using this cell lysate can show differences in protein half-life.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only