D-allo-Isoleucine, 97%
D-allo-Isoleucine, 97%
D-allo-Isoleucine, 97%
Thermo Scientific Chemicals

D-allo-Isoleucine, 97%

CAS: 1509-35-9 | C6H13NO2 | 131.18 g/mol
数量:
500 mg
1 g
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 357435000
価格(JPY)
-
数量:
500 mg
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1509-35-9
IUPAC Name(2R,3S)-2-amino-3-methylpentanoic acid
Molecular FormulaC6H13NO2
InChI KeyAGPKZVBTJJNPAG-CRCLSJGQSA-N
SMILESCC[C@H](C)[C@@H](N)C(O)=O
さらに表示
Appearance (Form)Crystalline powder or powder
Infrared spectrumConforms
Specific optical rotation-33° to -39° (20°C, 589 nm) (C=5, 1 N HCl)
Titration with HClO4>=96.0 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only