2-Chlorobenzenesulfonyl chloride, 98%
2-Chlorobenzenesulfonyl chloride, 98%
2-Chlorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

2-Chlorobenzenesulfonyl chloride, 98%

CAS: 2905-23-9 | C6H4Cl2O2S | 211.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 357460010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2905-23-9
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Clear yellow to orange-red
Appearance (Form)Liquid or solid
Refractive index1.5745 to 1.5775 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only