2,4-Difluorobenzenesulfonyl chloride, 98%
2,4-Difluorobenzenesulfonyl chloride, 98%
2,4-Difluorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

2,4-Difluorobenzenesulfonyl chloride, 98%

CAS: 13918-92-8 | C6H3ClF2O2S | 212.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 357490250
Price (USD)
210.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
210.00
Each
Chemical Identifiers
CAS13918-92-8
IUPAC Name2,4-difluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H3ClF2O2S
InChI KeyFJSAJUXIHJIAMD-UHFFFAOYSA-N
SMILESFC1=CC(F)=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration Argentometric>=97.5 % (ex Cl)
Refractive index1.5165 to 1.5185 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only