2-Bromo-3-methylpyridine, 95%
2-Bromo-3-methylpyridine, 95%
2-Bromo-3-methylpyridine, 95%
Thermo Scientific Chemicals

2-Bromo-3-methylpyridine, 95%

CAS: 3430-17-9 | C6H6BrN | 172.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3576600505 mLGlass bottle
Catalog number 357660050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3430-17-9
IUPAC Name2-bromo-3-methylpyridine
Molecular FormulaC6H6BrN
InChI KeyPZSISEFPCYMBDL-UHFFFAOYSA-N
SMILESCC1=CC=CN=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=94 %
Refractive index1.5650 to 1.5700 (20°C, 589 nm)
Appearance (Color)Clear light yellow to light brown
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only