2-Hydroxy-5-methylpyridine, 97%
2-Hydroxy-5-methylpyridine, 97%
2-Hydroxy-5-methylpyridine, 97%
Thermo Scientific Chemicals

2-Hydroxy-5-methylpyridine, 97%

CAS: 91914-06-6 | C6H7NO | 109.13 g/mol
Have Questions?
Catalog NumberQuantity
3576800505 g
Catalog number 357680050
Price (USD)
146.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
146.00
Each
Chemical Identifiers
CAS91914-06-6
IUPAC Name5-methyl-1,2-dihydropyridin-2-one
Molecular FormulaC6H7NO
InChI KeySOHMZGMHXUQHGE-UHFFFAOYSA-N
SMILESCC1=CNC(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Light yellow
Infrared spectrumConforms
GC>=96.0 %
Melting point180°C to 185°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only