Lithium trifluoromethanesulfonate, 98%
Lithium trifluoromethanesulfonate, 98%
Lithium trifluoromethanesulfonate, 98%
Lithium trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Lithium trifluoromethanesulfonate, 98%

CAS: 33454-82-9 | CF3LiO3S | 156.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 358710250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS33454-82-9
IUPAC Nametrifluoromethanesulfonic acid lithium
Molecular FormulaCHF3LiO3S
InChI KeyWDGKXRCNMKPDSD-UHFFFAOYSA-N
SMILES[Li].OS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Lithium (Li)4.34 to 4.56 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only