3-Bromophenylboronic acid, 97%
3-Bromophenylboronic acid, 97%
3-Bromophenylboronic acid, 97%
Thermo Scientific Chemicals

3-Bromophenylboronic acid, 97%

CAS: 89598-96-9 | C6H6BBrO2 | 200.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 358940050
Price (USD)
84.20
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
84.20
Each
Chemical Identifiers
CAS89598-96-9
IUPAC Name(3-bromophenyl)boronic acid
Molecular FormulaC6H6BBrO2
InChI KeyAFSSVCNPDKKSRR-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=96.0 %
Titration with NaOH96.0 to 110.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only