2-Formylphenylboronic acid, 98%
2-Formylphenylboronic acid, 98%
2-Formylphenylboronic acid, 98%
Thermo Scientific Chemicals

2-Formylphenylboronic acid, 98%

CAS: 40138-16-7 | C7H7BO3 | 149.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 359360010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS40138-16-7
IUPAC Name(2-formylphenyl)boronic acid
Molecular FormulaC7H7BO3
InChI KeyDGUWACLYDSWXRZ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink to orange
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only