Thermo Scientific Chemicals

4-(Methanesulfonyl)phenylboronic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 359510050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Methanesulfonyl)phenylboronic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 359510050
5 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
149104-88-1
IUPAC Name
(4-methanesulfonylphenyl)boronic acid
Molecular Formula
C7H9BO4S
InChI Key
VDUKDQTYMWUSAC-UHFFFAOYSA-N
SMILES
CS(=O)(=O)C1=CC=C(C=C1)B(O)O
Infrared spectrum
Conforms
HPLC
>=98.0 %
Appearance (Color)
Off-white
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text