Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
Acros Organics

Silica gel, for column chrom., ultrapure, 60-200 micron, 60A

CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 360060010
also known as 36006
HSN 28112200
Price (INR)
14,582.00
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (INR)
14,582.00
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Specific surface area470 to 530 m²/g
Pore diameter58 to 68 A
Pore volume0.70 to 0.85 ml/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
RUO – Research Use Only