Silica gel, functionalized, Amino-3, ca. 1.4mmol/g, part.size 40-63 micron
Silica gel, functionalized, Amino-3, ca. 1.4mmol/g, part.size 40-63 micron
Silica gel, functionalized, Amino-3, ca. 1.4mmol/g, part.size 40-63 micron
Thermo Scientific Chemicals

Silica gel, functionalized, Amino-3, ca. 1.4mmol/g, part.size 40-63 micron

CAS: 71888-97-6 | C9H23NO5Si2 | 281.455 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass Bottle
產品號碼 360180100
價格 (TWD)
-
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
化學識別
CAS71888-97-6
IUPAC Name(3-aminopropyl)triethoxysilane; silanedione
Molecular FormulaC9H23NO5Si2
InChI KeyJIBSXWZIQTVOKJ-UHFFFAOYSA-N
SMILESO=[Si]=O.CCO[Si](CCCN)(OCC)OCC
檢視更多
規格Specification Sheet規格表
Appearance (Form)Powder
Loading>=1.2 mmol/g (molecular)
Nitrogen (N)>=1.68 %
Appearance (Color)White to off-white
Particle size40 to 63 µm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesis
RUO – Research Use Only