1,1-Di(tert-butylperoxy)cyclohexane, 50% solution in mineral oil
1,1-Di(tert-butylperoxy)cyclohexane, 50% solution in mineral oil
1,1-Di(tert-butylperoxy)cyclohexane, 50% solution in mineral oil
Thermo Scientific Chemicals

1,1-Di(tert-butylperoxy)cyclohexane, 50% solution in mineral oil

CAS: 3006-86-8 | C14H28O4 | 260.374 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
250 mL
Catalog number 361310100
Price (USD)
-
Quantity:
10 mL
Request bulk or custom format
Chemical Identifiers
CAS3006-86-8
IUPAC Name1,1-bis(tert-butylperoxy)cyclohexane
Molecular FormulaC14H28O4
InChI KeyHSLFISVKRDQEBY-UHFFFAOYSA-N
SMILESCC(C)(C)OOC1(CCCCC1)OOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Hydroperoxides=<0.2 % (as TBHP)
Hydroperoxides=<0.2 % (as TBHP)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
RUO – Research Use Only