(S)-2-Isopropylsuccinic acid-1-methyl ester, 95%, 98% ee
(S)-2-Isopropylsuccinic acid-1-methyl ester, 95%, 98% ee
(S)-2-Isopropylsuccinic acid-1-methyl ester, 95%, 98% ee
Thermo Scientific Chemicals

(S)-2-Isopropylsuccinic acid-1-methyl ester, 95%, 98% ee

CAS: 208113-95-5 | C8H14O4 | 174.19 g/mol
Have Questions?
Catalog NumberQuantity
3621100101 g
Catalog number 362110010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS208113-95-5
IUPAC Name(3S)-3-(methoxycarbonyl)-4-methylpentanoate
Molecular FormulaC8H13O4
InChI KeyVHOGSWBAPDPWTG-LURJTMIESA-M
SMILESCOC(=O)[C@@H](CC([O-])=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Infrared spectrumConforms
Appearance (Form)Viscous liquid
GC>=94 %
Enantiomeric excess>=97.5 % (GC)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only