(S)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Thermo Scientific Chemicals
(S)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Thermo Scientific Chemicals
(S)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-3-Amino-3-phenylpropan-1-ol, 95%, 98% ee, Thermo Scientific Chemicals

CAS: 82769-76-4 | Molecular Formula: C9H13NO | Molecular Weight: 151.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
362172500250 mgGlass bottle
Catalog number 362172500
Price (USD)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS82769-76-4
IUPAC Name3-amino-3-phenylpropan-1-ol
Molecular FormulaC9H13NO
InChI KeySEQXIQNPMQTBGN-UHFFFAOYNA-N
SMILESNC(CCO)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Viscous liquid or low melting solid
GC>=94 %
Enantiomeric excess>=97.5 %
Appearance (Color)Colorless to white

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only