Nalgene™ Nonsterile PPCO Micro Packaging Vial Closures for 4.5mL Vials - FAQs
View additional product information for Nalgene™ Nonsterile PPCO Micro Packaging Vial Closures for 4.5mL Vials - FAQs (362826-0111)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.