(R)-N-BOC-Allylglycine, 95%, 98% ee
(R)-N-BOC-Allylglycine, 95%, 98% ee
(R)-N-BOC-Allylglycine, 95%, 98% ee
Thermo Scientific Chemicals

(R)-N-BOC-Allylglycine, 95%, 98% ee

CAS: 170899-08-8 | C10H17NO4 | 215.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Catalog number 362845000
Price (USD)
76.80
Each
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
76.80
Each
Chemical Identifiers
CAS170899-08-8
IUPAC Name(2R)-2-{[(tert-butoxy)carbonyl]amino}pent-4-enoate
Molecular FormulaC10H16NO4
InChI KeyBUPDPLXLAKNJMI-SSDOTTSWSA-M
SMILESCC(C)(C)OC(=O)N[C@H](CC=C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow
Appearance (Form)Viscous liquid or paste
Infrared spectrumConforms
Enantiomeric excess>=97.5 %
Specific optical rotation-9.5° to -12.5° (20°C, 589 nm) (c=1, methanol)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only