Hexaammineruthenium(III) chloride, 98%
Hexaammineruthenium(III) chloride, 98%
Hexaammineruthenium(III) chloride, 98%
Thermo Scientific Chemicals

Hexaammineruthenium(III) chloride, 98%

CAS: 14282-91-8 | Cl3H18N6Ru | 309.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 363340010
Price (USD)
161.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
161.00
Each
Chemical Identifiers
CAS14282-91-8
IUPAC Nameruthenium(3+) hexaamine trichloride
Molecular FormulaCl3H18N6Ru
InChI KeyGBDZMMXUOBAJMN-UHFFFAOYSA-K
SMILESN.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Elemental Analysis>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only