Nunc™ CryoTube™ Colored Caps
Nunc™ CryoTube™ Colored Caps
Nunc™ CryoTube™ Colored Caps
Nunc™ CryoTube™ Colored Caps
Thermo Scientific™
Nunc™ CryoTube™ Colored Caps
Color-code your sample vials with replacement polypropylene screw caps featuring a silicone gasket to provide the best seal for internally threaded cryotube vials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterility
363436ClearNon-sterile
336078ClearSterile
336081RedSterile
336088BlueSterile
336092YellowSterile
336094AssortedSterile
Catalog number 363436
Price (USD)
1,000.28
Online exclusive
1,167.25
Save 166.97
Case of 2500
Color:
Clear
Sterility:
Non-sterile
Price (USD)
1,000.28
Online exclusive
1,167.25
Save 166.97
Case of 2500
Color-code your sample vials. Thermo Scientific™ Nunc™ CryoTube™ Colored Caps are for use with Nunc internally threaded CryoTube vials and include a silicone gasket to provide the best seal. Nunc CryoTubes are the most widely used cryo vials in the world, stemming from the first cryo vials manufactured in the early 1960s. The well-proven design, the high certification level and the large portfolio of vials provide researchers with a choice of the vial best suited for their application and peace of mind that the sample can be retrieved viable and unharmed.
  • These replacement polypropylene screw caps have a silicone gasket to provide the best seal
  • Available nonsterile or irradiated
Specifications
Product TypeCap
ColorClear
MaterialPolypropylene
Product LineNunc
Quantity2500/Pk., 2500/Cs.
SterilityNon-sterile
Unit SizeCase of 2500