Bis(tricyclohexylphosphine)palladium(0), 98%
Bis(tricyclohexylphosphine)palladium(0), 98%
Bis(tricyclohexylphosphine)palladium(0), 98%
Thermo Scientific Chemicals

Bis(tricyclohexylphosphine)palladium(0), 98%

CAS: 33309-88-5 | C36H66P2Pd | 667.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
363511000100 mgGlass bottle
Catalog number 363511000
Price (USD)
-
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS33309-88-5
IUPAC Namebis(tricyclohexylphosphane) palladium
Molecular FormulaC36H66P2Pd
InChI KeyJGBZTJWQMWZVNX-UHFFFAOYSA-N
SMILES[Pd].C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light green
Appearance (Form)Powder
Infrared spectrumConforms
NMR (31P-NMR)>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only