Dichloro(1,5-cyclooctadiene)platinum(II), 98%
Dichloro(1,5-cyclooctadiene)platinum(II), 98%
Dichloro(1,5-cyclooctadiene)platinum(II), 98%
Thermo Scientific Chemicals

Dichloro(1,5-cyclooctadiene)platinum(II), 98%

CAS: 12080-32-9 | C8H12Cl2Pt | 374.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 363610010
Price (EUR)
236,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
236,00
Each
Chemical Identifiers
CAS12080-32-9
IUPAC Name(1Z,5Z)-cycloocta-1,5-diene; dichloroplatinum
Molecular FormulaC8H12Cl2Pt
InChI KeyVVAOPCKKNIUEEU-PHFPKPIQSA-L
SMILESCl[Pt]Cl.C1C\C=C/CC\C=C/1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Assay>=97.5 % (ex Pt)
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only