1,2-Bis(diphenylphosphino)benzene, 98%
1,2-Bis(diphenylphosphino)benzene, 98%
1,2-Bis(diphenylphosphino)benzene, 98%
Thermo Scientific Chemicals

1,2-Bis(diphenylphosphino)benzene, 98%

CAS: 13991-08-7 | C30H24P2 | 446.47 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3637500101 gGlass bottle
Catalog number 363750010
Price (USD)
70.65
Online exclusive
78.80
Save 8.15
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
70.65
Online exclusive
78.80
Save 8.15
Each
Chemical Identifiers
CAS13991-08-7
IUPAC Name[2-(diphenylphosphanyl)phenyl]diphenylphosphane
Molecular FormulaC30H24P2
InChI KeyNFRYVRNCDXULEX-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Assay>=97.5 %
Melting point184°C to 189°C
Appearance (Form)Crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only