1,1'-Bis(diisopropylphosphino)ferrocene, 98%, Thermo Scientific Chemicals
1,1'-Bis(diisopropylphosphino)ferrocene, 98%, Thermo Scientific Chemicals
1,1'-Bis(diisopropylphosphino)ferrocene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1'-Bis(diisopropylphosphino)ferrocene, 98%, Thermo Scientific Chemicals

CAS: 97239-80-0 | Molecular Formula: C22H36FeP2 | Molecular Weight: 418.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 363870025
Price (USD)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS97239-80-0
IUPAC Name1,1'-Bis(diisopropylphosphino)ferrocene
Molecular FormulaC22H36FeP2
SMILES[Fe].CC(C)P(C(C)C)c1cccc1.CC(C)P(C(C)C)c1cccc1
Molecular Weight (g/mol)418.32
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Appearance (Color)Yellow to orange
Infrared spectrumConforms
GC>=97.5 %
Melting point49°C to 53°C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only