Sodium tetraethylborate, 97%, pure, Thermo Scientific Chemicals
Sodium tetraethylborate, 97%, pure, Thermo Scientific Chemicals
Sodium tetraethylborate, 97%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium tetraethylborate, 97%, pure, Thermo Scientific Chemicals

Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 363880010
Price (USD)
81.70
Each
Request bulk or custom format
Price (USD)
81.70
Each
Chemical Identifiers
CAS15523-24-7
IUPAC Namesodium tetraethylboranuide
Molecular FormulaC8H20BNa
InChI KeySZSBMTRYJRHYNI-UHFFFAOYSA-N
SMILES[Na+].CC[B-](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=96.0 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only