Nalgene™ Break-Resistant Self-Zeroing Automatic Burets, 10mL - FAQs
Nalgene™ Break-Resistant Self-Zeroing Automatic Burets, 10mL - FAQs
View additional product information for Nalgene™ Break-Resistant Self-Zeroing Automatic Burets - FAQs (3640-0010PK, 3640-0010, 3640-0025PK, 3640-0050, 3640-0050PK, 3640-0025)
1 product FAQs found
What are your ASTM certified labware offerings?
Class B graduated cylinders meet ASTM E1272 and ISO 6706. Double-scale pharmaceutical graduates meet ASTM E1094. Class B volumetric flasks meet ASTM E288 and ISO 384. Burets meet ASTM E287. It is important to note that after autoclaving, even certified products may no longer be as accurate.