Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific™

Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
View all
Glass Bottle
Catalog number 364405000
Price (USD)
53.80
Each
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
53.80
Each
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=99.5 %
Appearance (Form)Liquid
Water=<0.005 % (Coulometric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only