Nalgene™ Self-Zeroing Buret Filler Kit
Nalgene™ Self-Zeroing Buret Filler Kit
Nalgene™ Self-Zeroing Buret Filler Kit
Nalgene™ Self-Zeroing Buret Filler Kit
Thermo Scientific™

Nalgene™ Self-Zeroing Buret Filler Kit

Save time and increase convenience. This filler kit converts any conventional glass or plastic 25mL or 50mL buret into an automatic-filling, self-zeroing buret.
Have Questions?
Catalog NumberFor Use With (Equipment)
3645-2550PK
also known as 3645-2550
25 or 50 mL Burette
Catalog number 3645-2550PK
also known as 3645-2550
Price (USD)
50.27
Online exclusive
55.90
Save 5.63
Each
For Use With (Equipment):
25 or 50 mL Burette
Price (USD)
50.27
Online exclusive
55.90
Save 5.63
Each

Save time and increase convenience. The Thermo Scientific™ Nalgene™ Self-Zeroing Buret Filler Kit converts any conventional glass or plastic 25mL or 50mL buret into an automatic-filling, self-zeroing buret. Avoid messy spills from manual fi

Operating Instructions:

The polypropylene fill tube connects to the end of the transfer tubing and is inserted into the mouth of the buret using the provided adapter. Adjust the fill tube tip so it is just above the zero mark of your buret. Squeeze the bottle to fill the buret above the zero mark. When you release the squeeze on the bottle, the excess titrant will be drawn back into the reservoir bottle leaving the titrant level in the buret at the zero line.

Includes:

  • 1000mL low-density polyethylene squeeze bottle
  • 4′ length (1.2m) of clear, flexible PVC tubing
  • Fill tube
  • Adapter
  • Closure for reagent bottle.
Buret not included.

Buret not included
Specifications
For Use With (Equipment)25 or 50 mL Burette
Includes1000 mL Squeeze Bottle, 4 ft. PVC Tubing, Fill Tube, Adapter and Closure
No. per Case4
No. per Pack1
MaterialLDPE
TypeBuret Filler Kit
Unit SizeEach