2-Methylresorcinol, 98%
2-Methylresorcinol, 98%
2-Methylresorcinol, 98%
Thermo Scientific Chemicals

2-Methylresorcinol, 98%

CAS: 608-25-3 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 364740250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS608-25-3
IUPAC Name2-methylbenzene-1,3-diol
Molecular FormulaC7H8O2
InChI KeyZTMADXFOCUXMJE-UHFFFAOYSA-N
SMILESCC1=C(O)C=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point118°C to 124°C
Appearance (Form)Crystalline powder
GC>=97.5 %
Appearance (Color)Beige to pink to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only