Potassium tert-butoxide, pure, 20 wt.% solution in THF
Potassium tert-butoxide, pure, 20 wt.% solution in THF
Potassium tert-butoxide, pure, 20 wt.% solution in THF
Thermo Scientific Chemicals

Potassium tert-butoxide, pure, 20 wt.% solution in THF

CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 364990500
Price (USD)
67.65
Online Exclusive
75.40
Save 7.75 (10%)
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
67.65
Online Exclusive
75.40
Save 7.75 (10%)
Each
Chemical Identifiers
CAS865-47-4
IUPAC Namepotassium 2-methylpropan-2-olate
Molecular FormulaC4H9KO
InChI KeyLPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES[K+].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to amber
Appearance (Form)Solution
Strength19 to 21 % (Acidimetric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only