HATU, 97%
Thermo Scientific Chemicals

HATU, 97%

CAS: 148893-10-1 | C10H15F6N6OP | 380.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 365310250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS148893-10-1
IUPAC Name[(dimethylamino)({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})methylidene]dimethylazanium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC10H15F6N6OP
InChI KeyJNWBBCNCSMBKNE-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=CN=C12)=[N+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder and/or lumps
Infrared spectrumConforms
Melting point183°C to 193°C (Dec)
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only