3-(Trifluoromethyl)phenethylamine, 98%
3-(Trifluoromethyl)phenethylamine, 98%
3-(Trifluoromethyl)phenethylamine, 98%
Thermo Scientific Chemicals

3-(Trifluoromethyl)phenethylamine, 98%

CAS: 52516-30-0 | C9H10F3N | 189.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 365520010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52516-30-0
IUPAC Name2-[3-(trifluoromethyl)phenyl]ethan-1-aminium
Molecular FormulaC9H11F3N
InChI KeyBPVYCXMGJPKOTQ-UHFFFAOYSA-O
SMILES[NH3+]CCC1=CC=CC(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
Refractive index1.4635 to 1.4655 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only