Clear Round-Bottom Immuno Nonsterile 96-Well Plates
Clear Round-Bottom Immuno Nonsterile 96-Well Plates
Clear Round-Bottom Immuno Nonsterile 96-Well Plates
Clear Round-Bottom Immuno Nonsterile 96-Well Plates
Thermo Scientific™
Clear Round-Bottom Immuno Nonsterile 96-Well Plates
Carry out colormetric assays with reliable and reproducible results using these clear, round-bottom, immuno nonsterile 96-well plates, offered with a choice of surface.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface Treatment
3655TS
also known as 3655
Immulon 2 HB
3555TS
also known as 3555
Immulon 1 B
449824MaxiSorp
475434PolySorp
Catalog number 3655TS
also known as 3655
Price (USD)
235.68
Online exclusive
275.07
Save 39.39
Case of 50
Surface Treatment:
Immulon 2 HB
Price (USD)
235.68
Online exclusive
275.07
Save 39.39
Case of 50
Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates, offered with a choice of surface for optimal binding. Features Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
  • Clear polystyrene
  • Choice of a variety of surfaces for optimal binding
  • Alphanumerically labeled rows and columns
Surfaces:
  • Thermo Scientific PolySorp and Immulon 1B are for adsorption of hydrophobic molecules.
  • Thermo Scientific Immulon 2HB is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
Ordering Information:
Please note that availability may vary by country.
Specifications
Binding PropertySlightly Hydrophilic
Detection MethodColorimetric
FormatStandard
No. per Case50
No. per Pack10
Packaging50 Cases
Plate BlockingNone
Product TypeMicroplate
Target MoleculeProteins
Volume (Metric) Well280 μL
Volume (Metric) Working180 μL
Array8 x 12
BarcodeNo Barcode
ColorClear
MaterialPolystyrene
No. of Wells96
SterilityNon-sterile
Surface TreatmentImmulon 2 HB
Well ShapeRound
Unit SizeCase of 50
Frequently asked questions (FAQs)
For Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, is the surface treatment solely on the bottom of the well, or is it also applied to the sides?
For energy treated plates, the entire plate surface receives the energy treatment. The MaxiSorp, PolySorp, MediSorp and MultiSorp plates are energy treated plates. For traditionally treated plates, the well surface is treated up to the listed working volume.