Methyl 4-bromobutyrate, 98+%
Methyl 4-bromobutyrate, 98+%
Methyl 4-bromobutyrate, 98+%
Thermo Scientific Chemicals

Methyl 4-bromobutyrate, 98+%

CAS: 4897-84-1 | C5H9BrO2 | 181.03 g/mol
数量:
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 365830100
価格(JPY)
-
数量:
10 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4897-84-1
IUPAC Namemethyl 4-bromobutanoate
Molecular FormulaC5H9BrO2
InChI KeyQAWFLJGZSZIZHO-UHFFFAOYSA-N
SMILESCOC(=O)CCCBr
さらに表示
GC>=98.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4610 to 1.4630 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only